COM497523258 Combi-Blocks, Inc.

Benzyl 3-((methylsulfonyl)oxy)pyrrolidine-1-carboxylate

  • MDL number MFCD16620796
  • Cas Number : 138232-44-7
  • Molecular Weight : 299.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3384
Availability details for
Quantity
  • Availability
  • Add to Cart