COM497523276 Combi-Blocks, Inc.

5-Bromo-2,4-difluoro-3-methylbenzenamine

  • MDL number MFCD16620248
  • Cas Number : 112822-79-4
  • Molecular Weight : 222.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3403

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3403
Availability details for
Quantity
  • Availability
  • Add to Cart