COM497523298 Combi-Blocks, Inc.

tert-Butyl 3-[(benzylideneamino)methyl] piperidine-1-carboxylate

  • MDL number MFCD16620824
  • Cas Number : 1824819-67-1
  • Molecular Weight : 302.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3425
Availability details for
Quantity
  • Availability
  • Add to Cart