COM497523312 Combi-Blocks, Inc.

tert-Butyl 4-cyanobenzoate

  • MDL number MFCD11041298
  • Cas Number : 55696-50-9
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3439
Availability details for
Quantity
  • Availability
  • Add to Cart