COM497523330 Combi-Blocks, Inc.

tert-Butyl 5-fluoroisoindoline-2-carboxylate

  • MDL number MFCD11878365
  • Cas Number : 871013-94-4
  • Molecular Weight : 237.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3458

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3458
Availability details for
Quantity
  • Availability
  • Add to Cart