COM497523344 Combi-Blocks, Inc.

2-tert-Butyl 5-methyl isoindoline-2,5-dicarboxylate

  • MDL number MFCD16620899
  • Cas Number : 368441-44-5
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3473
Availability details for
Quantity
  • Availability
  • Add to Cart