COM497523391 Combi-Blocks, Inc.

[(4',4'-Ethylenedioxy)cyclohexyl]methyl methanesulfonate

  • MDL number MFCD16621693
  • Cas Number : 147767-55-3
  • Molecular Weight : 250.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3522
Availability details for
Quantity
  • Availability
  • Add to Cart