COM497523399 Combi-Blocks, Inc.

Ethoxycarbonylamino-acetic acid methyl ester

  • MDL number MFCD09033273
  • Cas Number : 13756-47-3
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3530
Availability details for
Quantity
  • Availability
  • Add to Cart