COM497523499 Combi-Blocks, Inc.

2-(1-Benzylpiperidin-3-yl)pyridine

  • MDL number MFCD14582905
  • Cas Number : 861907-77-9
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3634
Availability details for
Quantity
  • Availability
  • Add to Cart