COM497523507 Combi-Blocks, Inc.

1,3-Bis(5-methylpyridin-2-yl)urea

  • MDL number MFCD14280313
  • Cas Number : 63272-29-7
  • Molecular Weight : 242.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3642

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3642
Availability details for
Quantity
  • Availability
  • Add to Cart