COM497523520 Combi-Blocks, Inc.

2-(4-(tert-Butoxycarbonyl(methyl)amino) phenyl)acetic acid

  • MDL number MFCD14636446
  • Cas Number : 251643-18-2
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3656
Availability details for
Quantity
  • Availability
  • Add to Cart