COM497523523 Combi-Blocks, Inc.

tert-Butyl 1-(1-((benzyloxy)carbonyl) pyrrolidin-3-yl)piperidin-4-ylcarbamate

  • MDL number MFCD14635668
  • Cas Number : 1823288-48-7
  • Molecular Weight : 403.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3660

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3660
Availability details for
Quantity
  • Availability
  • Add to Cart