COM497523538 Combi-Blocks, Inc.

Di-tert-butyl 1,2-diazepane-1,2-dicarboxylate

  • MDL number MFCD14635704
  • Cas Number : 939964-15-5
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3675

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3675
Availability details for
Quantity
  • Availability
  • Add to Cart