COM497523539 Combi-Blocks, Inc.

tert-Butyl 3-((ethoxycarbonyl)methoxy)piperidine-1-carboxylate

  • MDL number MFCD14635717
  • Cas Number : 1824091-88-4
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3676

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3676
Availability details for
Quantity
  • Availability
  • Add to Cart