COM497523544 Combi-Blocks, Inc.

2-Hydroxymethyl-indole-1-carboxylic acid tert-butyl ester

  • MDL number MFCD09032312
  • Cas Number : 174180-53-1
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3681
Availability details for
Quantity
  • Availability
  • Add to Cart