COM497523620 Combi-Blocks, Inc.

Ethyl 2-(4-nitrophenoxy)-2-methylpropanoate

  • MDL number MFCD03900496
  • Cas Number : 23501-39-5
  • Molecular Weight : 253.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3761

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3761
Availability details for
Quantity
  • Availability
  • Add to Cart