COM497523645 Combi-Blocks, Inc.

6-(Methoxycarbonyl)-2-naphthoic acid

  • MDL number MFCD00805122
  • Cas Number : 7568-08-3
  • Molecular Weight : 230.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3787

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3787
Availability details for
Quantity
  • Availability
  • Add to Cart