COM497523692 Combi-Blocks, Inc.

2-(Formylamino)pyridine

  • MDL number MFCD00043008
  • Cas Number : 34813-97-3
  • Molecular Weight : 122.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3835
Availability details for
Quantity
  • Availability
  • Add to Cart