COM497523695 Combi-Blocks, Inc.

4-((R)-Pyrrolidin-3-yloxy)-pyridine hydrochloride

  • MDL number MFCD18380459
  • Cas Number : 1264033-82-0
  • Molecular Weight : 200.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3838

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3838
Availability details for
Quantity
  • Availability
  • Add to Cart