COM497523809 Combi-Blocks, Inc.

6-Nitroisochroman

  • MDL number MFCD08234693
  • Cas Number : 207804-97-5
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4013

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4013
Availability details for
Quantity
  • Availability
  • Add to Cart