COM497523826 Combi-Blocks, Inc.

4-Fluoro-n-methyl-3-(trifluoromethyl)aniline

  • MDL number MFCD11655738
  • Cas Number : 1187386-25-9
  • Molecular Weight : 193.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4053
Availability details for
Quantity
  • Availability
  • Add to Cart