COM497523891 Combi-Blocks, Inc.

1-Bromo-5-methoxy-4-methyl-2-nitrobenzene

  • MDL number MFCD28291981
  • Cas Number : 1807224-98-1
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4141
Availability details for
Quantity
  • Availability
  • Add to Cart