COM497523938 Combi-Blocks, Inc.

2-Cyano-5-fluorobenzyl chloride

  • MDL number MFCD18394047
  • Cas Number : 1261742-22-6
  • Molecular Weight : 169.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4201
Availability details for
Quantity
  • Availability
  • Add to Cart