COM497523939 Combi-Blocks, Inc.

1-Bromo-3-(bromomethyl)-2-chlorobenzene

  • MDL number MFCD18390317
  • Cas Number : 1044256-89-4
  • Molecular Weight : 284.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4202

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4202
Availability details for
Quantity
  • Availability
  • Add to Cart