COM497523943 Combi-Blocks, Inc.

5-Chloro-2,4-difluorophenylacetonitrile

  • MDL number MFCD28402683
  • Cas Number : 1429422-26-3
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4206

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4206
Availability details for
Quantity
  • Availability
  • Add to Cart