COM497524273 Combi-Blocks, Inc.

1-(1-Methyl-1-phenylethyl)piperidine

  • MDL number MFCD00870884
  • Cas Number : 92321-29-4
  • Molecular Weight : 203.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4646
Availability details for
Quantity
  • Availability
  • Add to Cart