COM497524375 Combi-Blocks, Inc.

6-(Trifluoromethoxy)pyridin-2-amine

  • MDL number MFCD20542914
  • Cas Number : 1131007-45-8
  • Molecular Weight : 178.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-4753
Availability details for
Quantity
  • Availability
  • Add to Cart