COM497524707 Combi-Blocks, Inc.

4-Chloroisoindoline

  • MDL number MFCD08234860
  • Cas Number : 123594-04-7
  • Molecular Weight : 153.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5105
Availability details for
Quantity
  • Availability
  • Add to Cart