COM497524873 Combi-Blocks, Inc.

Boc-d-his(3-bom)-oh

  • MDL number MFCD00076940
  • Cas Number : 99310-01-7
  • Molecular Weight : 375.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5277
Availability details for
Quantity
  • Availability
  • Add to Cart