COM497524882 Combi-Blocks, Inc.

(2S,4S)-1-((9H-Fluoren-9-yl)methyl) 2-benzyl 4-hydroxypyrrolidine-1,2-dicarboxylate

  • MDL number MFCD23135321
  • Cas Number : 439290-35-4
  • Molecular Weight : 443.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5286

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5286
Availability details for
Quantity
  • Availability
  • Add to Cart