COM497525038 Combi-Blocks, Inc.

Cis-3-hexenyl salicylate

  • MDL number MFCD00036486
  • Cas Number : 65405-77-8
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5499
Availability details for
Quantity
  • Availability
  • Add to Cart