COM497525122 Combi-Blocks, Inc.

Ethyl 4-(trifluoromethyl)-1h-indole-2-carboxylate

  • MDL number MFCD09954812
  • Cas Number : 317-60-2
  • Molecular Weight : 257.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5847

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-5847
Availability details for
Quantity
  • Availability
  • Add to Cart