COM497525212 Combi-Blocks, Inc.

Chloromethyl(dimethyl)methoxysilane

  • MDL number MFCD05664349
  • Cas Number : 18143-33-4
  • Molecular Weight : 138.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6168

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6168
Availability details for
Quantity
  • Availability
  • Add to Cart