COM497525298 Combi-Blocks, Inc.

(2R)-2-(2-Methylphenyl)pyrrolidine

  • MDL number MFCD06762523
  • Cas Number : 914299-83-5
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6415
Availability details for
Quantity
  • Availability
  • Add to Cart