COM497525380 Combi-Blocks, Inc.

3-(Aminomethyl)-6-methyl-4-(trifluoromethyl)-2(1h)-pyridinone

  • MDL number MFCD06212992
  • Cas Number : 771581-72-7
  • Molecular Weight : 206.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6581

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6581
Availability details for
Quantity
  • Availability
  • Add to Cart