COM497525415 Combi-Blocks, Inc.

(2-Methoxypyrimidin-5-yl)methanamine

  • MDL number MFCD08667859
  • Cas Number : 944899-18-7
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6655

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6655
Availability details for
Quantity
  • Availability
  • Add to Cart