COM497525419 Combi-Blocks, Inc.

(1S,3S,5S)-2-Azabicyclo[3.1.0]hexane-3-carboxylic acid

  • MDL number MFCD08691410
  • Cas Number : 214193-13-2
  • Molecular Weight : 127.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6662

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6662
Availability details for
Quantity
  • Availability
  • Add to Cart