COM497525596 Combi-Blocks, Inc.

tert-Butyl 3-oxo-1,2,3,5-tetrahydrospiro[benzo[c]azepine-4,4'-piperidine]-1'-carboxylate

  • MDL number MFCD12198647
  • Cas Number : 1160247-91-5
  • Molecular Weight : 330.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6980
Availability details for
Quantity
  • Availability
  • Add to Cart