COM497525683 Combi-Blocks, Inc.

Ethyl 3-(4-hydroxypiperidin-1-yl)-3-methylpyrrolidine-1-carboxylate

  • MDL number MFCD13659309
  • Cas Number : 1131451-63-2
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7129

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7129
Availability details for
Quantity
  • Availability
  • Add to Cart