COM497525696 Combi-Blocks, Inc.

7-Oxa-2-azaspiro[4.5]decane

  • MDL number MFCD14581264
  • Cas Number : 72088-42-7
  • Molecular Weight : 141.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7153
Availability details for
Quantity
  • Availability
  • Add to Cart