COM497525896 Combi-Blocks, Inc.

1-(1-Methyl-1h-imidazol-4-yl)-ethanol

  • MDL number MFCD18432780
  • Cas Number : 70702-82-8
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7532
Availability details for
Quantity
  • Availability
  • Add to Cart