COM497525985 Combi-Blocks, Inc.

5-(Methylsulfonyl)-7-azaindole

  • MDL number MFCD20259032
  • Cas Number : 1036383-51-3
  • Molecular Weight : 196.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7761

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7761
Availability details for
Quantity
  • Availability
  • Add to Cart