COM497526498 Combi-Blocks, Inc.

3-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-([2-(trimethylsilyl)ethoxy]methyl)-1h-pyrrolo[2,3-b]pyridine-5-carbonitrile

  • MDL number MFCD27987788
  • Cas Number : 1429906-71-7
  • Molecular Weight : 399.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9103

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9103
Availability details for
Quantity
  • Availability
  • Add to Cart