COM497526664 Combi-Blocks, Inc.

2,2,2-Trifluoro-1-(pyridin-3-yl)ethanone hcl h2o

  • MDL number MFCD28405557
  • Cas Number : 33284-21-8
  • Molecular Weight : 229.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9655

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9655
Availability details for
Quantity
  • Availability
  • Add to Cart