COM497526697 Combi-Blocks, Inc.

(2S,4S)-1-(tert-Butoxycarbonyl)-4-cyanopyrrolidine-2-carboxylic acid

  • MDL number MFCD14582677
  • Cas Number : 132622-71-0
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9727
Availability details for
Quantity
  • Availability
  • Add to Cart