COM497526786 Combi-Blocks, Inc.

(3R,4R)-1-Benzyl-3,4-dihydroxypyrrolidine-2,5-dione

  • MDL number MFCD03695453
  • Cas Number : 75172-31-5
  • Molecular Weight : 221.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9875

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-9875
Availability details for
Quantity
  • Availability
  • Add to Cart