COM497526876 Combi-Blocks, Inc.

1,2,3,4-Tetrahydropyrazino[2,1-a]isoindol-6(10bh)-one

  • MDL number MFCD12198737
  • Cas Number : 79016-59-4
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3850

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-3850
Availability details for
Quantity
  • Availability
  • Add to Cart