COM497526905 Combi-Blocks, Inc.

Trans-tert-butyl-4-oxohexahydro-1h-pyrrolo[3,4-c]pyridine-2(3h)-carboxylate

  • MDL number MFCD12198711
  • Cas Number : 1251012-56-2
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5099
Availability details for
Quantity
  • Availability
  • Add to Cart