COM497526932 Combi-Blocks, Inc.

tert-Butyl 2-chloro-4-methyl-7,8-dihydro-1,6-naphthyridine-6(5h)-carboxylate

  • MDL number MFCD27997374
  • Cas Number : 1421254-01-4
  • Molecular Weight : 282.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6265

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-6265
Availability details for
Quantity
  • Availability
  • Add to Cart