COM497527057 Combi-Blocks, Inc.

1-[(1-Aminopropan-2-yl)oxy]-2-chlorobenzene, HCl

  • MDL number MFCD02684120
  • Cas Number : 1021871-58-8
  • Molecular Weight : 222.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1650

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1650
Availability details for
Quantity
  • Availability
  • Add to Cart